Synonyms:
Basic Green 4Bis (N-{4-[[4-(dimethylamino)phenyl](phenyl)methylene]cyclohexa-2,5-dien-1-ylidene}-N-methylmethanaminium) trioxalateN,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate
A5100-1G
TCA5100-1GEA
81.5
USD
InStock
TCA5100-1G
Malachite green oxalate ≥95.0% (by titrimetric analysis)
Malachite green oxalate
[Sensitive spectrophotometric reagent for PO4]
CAS Number: 2437-29-8
MDL Number: MFCD00011766
Molecular Formula: C52H54N4O12
Molecular Weight: 927.02
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 164
Lambda max.: 617 nm (H2O)
Product Categories: Bioscience, Stains & Dyes, Analytical Science, Ion Association Reagents
Other Product Names / Synonyms: Basic Green 4