Synonyms:
2-Bromo-1-(3-methoxyphenyl)ethan-1-one 2-Bromo-3'-methoxyacetophenone 3-Methoxyphenacyl bromide Bromomethyl m-methoxyphenyl ketone α-Bromo-3-methoxyacetophenone
A5505-5G
TCA5505-5GEA
110.64
USD
InStock
TCA5505-5G
ω-Bromo-3-methoxyacetophenone ≥99.0% (by titrimetric analysis)
ω-Bromo-3-methoxyacetophenone
[for HPLC Labeling]
CAS Number: 5000-65-7
MDL Number: MFCD00000199
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 63
Storage Temperature: 0-10°C
Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 3-(Bromoacetyl)anisole, alpha-Bromo-3'-methoxyacetophenone