Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dinitrobenzoyl chloride ≥99.0% (by titrimetric analysis)

Supplier: TCI America

A5511-5G
TCA5511-5GEA 48.72 USD InStock
TCA5511-5G
3,5-Dinitrobenzoyl chloride ≥99.0% (by titrimetric analysis)
3,5-Dinitrobenzoyl chloride
[for HPLC Labeling]
CAS Number: 99-33-2
MDL Number: MFCD00007248
Molecular Formula: C7H3ClN2O5
Molecular Weight: 230.56
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Boiling point (°C): 201
Melting point (°C): 70


Formula: C₇H₃ClN₂O₅
MW: 230.56 g/mol
Boiling Pt: 201 °C
Melting Pt: 70 °C
MDL Number: MFCD00007248
CAS Number: 99-33-2
UN: 3261
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/13/28932113.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty