Synonyms:
Trimethylsilyl N-trimethylsilylacetamidate BSA BSA DerivatisationSilyl Derivatisation
A5601-5ML
TCA5601-5MLEA
33.5
USD
InStock
TCA5601-5ML
N,O-Bis(trimethylsilyl)acetamide ≥80.0% (by GC) for gas chromatography
N,O-Bis(trimethylsilyl)acetamide
Form: Clear Liquid
Boiling point (°C): 73
Melting point (°C): -24
Flash Point (°C): 42
Product Categories: Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: N,O-Bis(trimethylsilyl)acetamide