Synonyms:
Trimethylsilyl N-trimethylsilylacetamidate BSA BSA DerivatisationSilyl Derivatisation
A5602-5ML
TCA5602-5MLEA
77
USD
InStock
TCA5602-5ML
N,O-Bis(trimethylsilyl)acetamide 25% in acetonitrile (TMS-BA) for gas chromatography
N,O-Bis(trimethylsilyl)acetamide
[for Gas Chromatography]
CAS Number: 10416-59-8
MDL Number: MFCD00008270
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 10
Storage Temperature: 0-10°C
Other Product Names / Synonyms: N,O-Bis(trimethylsilyl)acetamide, BSA