Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trifluoroacetic acid ∼0.5 M in water

Supplier: TCI America
Synonyms: TFA

A5711-10ML
TCA5711-10MLEA 36.84 USD InStock
TCA5711-10ML
Trifluoroacetic acid ∼0.5 M in water
Trifluoroacetic acid
[Ion-Pair Reagent for LC-MS]
CAS Number: 76-05-1
MDL Number: MFCD00004169
Molecular Formula: C2HF3O2
Molecular Weight: 114.02
Form: Clear Liquid
Color: Colorless

Other Product Names / Synonyms: IPC-PFFA-2, TFA


Formula: CF₃COOH
MW: 114.02 g/mol
Density: 1.490 g/cm³ (20 °C)
MDL Number: MFCD00004169
CAS Number: 76-05-1
UN: 2699
ADR: 8,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/69/28932169.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty