Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,O-Bis(trimethylsilyl)acetamide 25% in acetonitrile

Supplier: TCI America
Synonyms: Trimethylsilyl N-trimethylsilylacetamidate BSA BSA DerivatisationSilyl Derivatisation

B0510-12ML
TCB0510-12MLEA 64.96 USD InStock
TCB0510-12ML
N,O-Bis(trimethylsilyl)acetamide 25% in acetonitrile
N,O-Bis(trimethylsilyl)acetamide
[Trimethylsilylating Reagent, for NH2 compounds]
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 10
Storage Temperature: 0-10°C

Other Product Names / Synonyms: TMS-BA, BSA


Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
Flash Pt: 10 °C
MDL Number: MFCD00008270
CAS Number: 10416-59-8
UN: 2920
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/24/62/28932462.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty