Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

N,O-Bis(trimethylsilyl)acetamide kit ≥80% (by GC)

Supplier: TCI America
Synonyms: BSASilyl DerivatisationTrimethylsilyl N-(trimethylsilyl)acetimidateTrimethylsilyl N-trimethylsilylacetamidate

B0911-1KIT
TCB0911-KITEA 102 USD InStock
TCB0911-KIT
N,O-Bis(trimethylsilyl)acetamide kit ≥80% (by GC)
N,O-Bis(trimethylsilyl)acetamide
Kit BSA 1 mL × 8 / Reaction vial, capacity 2 mL × 8


Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
MDL Number: MFCD00008270
CAS Number: 10416-59-8
UN: 2920
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/27/94/28932794.jpg
Order Now
Specification Test Results
Purity/Analysis Method 75.0% (GC)
Boiling point (°C) 73
Melting point (°C) -24
Flash Point (°C) 42
Specific Gravity (20/20) 0.83
Storage Temperature 0-10°C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty