Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture) ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: 1,4-Di(aminomethyl)cyclohexaneHexahydro-p-xylylenediamine

B1083-25G
TCB1083-25GEA 193.57 USD InStock
TCB1083-25G
1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture) ≥98.0% (by GC, titration analysis)
1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture)
CAS Number: 2549-93-1
MDL Number: MFCD00039463
Molecular Formula: C8H18N2
Molecular Weight: 142.25
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 106
Specific Gravity (20/20): 0.95

Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 1,4-Di(aminomethyl)cyclohexane, Hexahydro-p-xylylenediamine


Formula: C₈H₁₈N₂
MW: 142.25 g/mol
Density: 0.95 (20 °C)
Flash Pt: 106 °C
MDL Number: MFCD00039463
CAS Number: 2549-93-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/29/38/28932938.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty