Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-fluorotoluene ≥98.0% (by GC)

Supplier: TCI America
Synonyms: 1-Bromo-3-fluoro-2-methylbenzene

B2392-1G
TCB2392-1GEA 55 USD InStock
TCB2392-1G
2-Bromo-6-fluorotoluene ≥98.0% (by GC)
2-Bromo-6-fluorotoluene
CAS Number: 1422-54-4
MDL Number: MFCD00051438
Molecular Formula: C7H6BrF
Molecular Weight: 189.03
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 76
Specific Gravity (20/20): 1.53

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Bromo-3-fluoro-2-methylbenzene


Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 180 °C
Density: 1.53 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00051438
CAS Number: 1422-54-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/41/29/28934129.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty