Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromobenzenediazonium tetrafluoroborate ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

B3114-5G
TCB3114-5GEA 110.09 USD InStock
TCB3114-5G
4-Bromobenzenediazonium tetrafluoroborate ≥97.0% (by HPLC, titration analysis)
4-Bromobenzenediazonium tetrafluoroborate
CAS Number: 673-40-5
MDL Number: MFCD00011894
Molecular Formula: C6H4BBrF4N2
Molecular Weight: 270.82
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 133


Formula: C₆H₄BBrF₄N₂
Melting Pt: 133 °C
MDL Number: MFCD00011894
CAS Number: 673-40-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/45/54/28934554.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty