Synonyms:
gamma-Benzyl L-glutamate
, 5-Benzyl L-Glutamate
, H-Glu(OBzl)-OH
, γ-Benzyl-L-Glutamate
B3316-5G
TCB3316-5GEA
38
USD
InStock
TCB3316-5G
L-Glutamic acid 5-benzyl ester ≥98.0% (by HPLC, titration analysis)
L-Glutamic acid 5-benzyl ester
CAS Number: 1676-73-9
MDL Number: MFCD00002633
Molecular Formula: C12H15NO4
Molecular Weight: 237.26
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 158
Specific rotation [a]20/D: 18 deg (C=1, AcOH)
Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: L-Glutamic Acid 5-Benzyl Ester, H-Glu(OBzl)-OH