Synonyms:
1-(Phenylmethyl)-3-pyrrolidinone
, 1-Benzylpyrrolidin-3-one
B3606-5G
B3606-25G
TCB3606-5GEA
141.11
USD
InStock
TCB3606-5G
TCB3606-25G
1-Benzyl-3-pyrrolidinone ≥98.0% (by GC, titration analysis)
1-Benzyl-3-pyrrolidinone
CAS Number: 775-16-6
MDL Number: MFCD00005342
Molecular Formula: C11H13NO
Molecular Weight: 175.23
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 109
Flash Point (°C): 110
Specific Gravity (20/20): 1.07
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1-Benzyl-3-pyrrolidinone