Synonyms:
L-Glutamic Acid 5-tert-Butyl 1-Methyl Ester HydrochlorideH-Glu(OtBu)-OMe.HCl
B3670-1G
B3670-5G
TCB3670-5GEA
186
USD
InStock
TCB3670-5G
TCB3670-1G
5-tert-Butyl-1-methyl-L-glutamate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
5-tert-Butyl-1-methyl-L-glutamate hydrochloride
CAS Number: 6234-01-1
MDL Number: MFCD00153438
Molecular Formula: C10H19NO4
Molecular Weight: 253.72
Purity/Analysis Method: 98.0% (N,T)
Form: Crystal
Melting point (°C): 128
Specific rotation [a]20/D: 26 deg (C=1, MeOH)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: L-Glutamic Acid 5-tert-Butyl 1-Methyl Ester Hydrochloride, H-Glu(OtBu)-OMe.HCl