Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Bromoheptanoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 7-Bromoenanthic Acid

B3671-5G
TCB3671-5GEA 179.57 USD InStock
TCB3671-5G
7-Bromoheptanoic acid ≥98.0% (by titrimetric analysis)
7-Bromoheptanoic acid
CAS Number: 30515-28-7
MDL Number: MFCD00037800
Molecular Formula: C7H13BrO2
Molecular Weight: 209.08
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Boiling point (°C): 166
Freezing point (°C): 27

Other Product Names / Synonyms: 7-Bromoenanthic Acid


Formula: C₇H₁₃BrO₂
MW: 209.08 g/mol
Boiling Pt: 166 °C
MDL Number: MFCD00037800
CAS Number: 30515-28-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/49/90/28934990.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty