Synonyms:
6-Bromopyridine-2-carboxaldehyde
, 6-Bromopyridine-2-carbaldehyde
, 6-Bromo-2-pyridinecarboxaldehyde
B3716-1G
B3716-5G
TCB3716-5GEA
116.6
USD
InStock
TCB3716-5G
TCB3716-1G
2-Bromo-6-formylpyridine ≥98.0% (by GC)
2-Bromo-6-formylpyridine
CAS Number: 34160-40-2
MDL Number: MFCD02683546
Molecular Formula: C6H4BrNO
Molecular Weight: 186.01
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 81
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2-Bromo-6-formylpyridine, 6-Bromopicolinaldehyde