Synonyms:
2-Bromopropionylchloride
B4107-100G
B4107-25G
TCB4107-25GEA
83.49
USD
InStock
TCB4107-25G
TCB4107-100G
(±)-2-Bromopropionyl chloride ≥97.0% (by titrimetric analysis)
(±)-2-Bromopropionyl chloride
CAS Number: 7148-74-5
MDL Number: MFCD00000711
Molecular Formula: C3H4BrClO
Molecular Weight: 171.42
Purity/Analysis Method: 97.0% (T)
Form: Clear Liquid
Boiling point (°C): 131
Flash Point (°C): 51
Specific Gravity (20/20): 1.67