Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Butyl-3-[[2'-cyano-[1,1'-biphenyl]-4-yl]methyl]-1,3-diazaspiro[4,4]non-1-en-4-one ≥97.0% (by total nitrogen basis)

Supplier: TCI America
Synonyms: 4'-[(2-Butyl-4-oxo-1,3-diazaspiro[4,4]non-1-en-3-yl)methyl]-[1,1'-biphenyl]-2-carbonitrile

B4143-1G B4143-5G
TCB4143-1GEA 79.14 USD InStock
TCB4143-1G TCB4143-5G
2-Butyl-3-[[2'-cyano-[1,1'-biphenyl]-4-yl]methyl]-1,3-diazaspiro[4,4]non-1-en-4-one ≥97.0% (by total nitrogen basis)
2-Butyl-3-[[2'-cyano-[1,1'-biphenyl]-4-yl]methyl]-1,3-diazaspiro[4,4]non-1-en-4-one
CAS Number: 138401-24-8
MDL Number: MFCD06658242
Molecular Formula: C25H27N3O
Molecular Weight: 385.51
Purity/Analysis Method: 97.0% (N)
Form: Crystal
Melting point (°C): 93

Other Product Names / Synonyms: 4'-[(2-Butyl-4-oxo-1,3-diazaspiro[4,4]non-1-en-3-yl)methyl]-[1,1'-biphenyl]-2-carbonitrile


Formula: C₂₅H₂₇N₃O
MW: 385.51 g/mol
Melting Pt: 93 °C
MDL Number: MFCD06658242
CAS Number: 138401-24-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/54/67/28935467.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty