Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bifonazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: 1-(alpha,4-Diphenylbenzyl)imidazole

B4173-5G
TCB4173-5GEA 103.53 USD InStock
TCB4173-5G
Bifonazole ≥98.0% (by GC, titration analysis)
Bifonazole
CAS Number: 60628-96-8
MDL Number: MFCD00865567
Molecular Formula: C22H18N2
Molecular Weight: 310.40
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 149

Product Categories: Bioscience, Antifungal Ingredients
Other Product Names / Synonyms: 1-(alpha,4-Diphenylbenzyl)imidazole


Formula: C₂₂H₁₈N₂
MW: 310.40 g/mol
Melting Pt: 149 °C
MDL Number: MFCD00865567
CAS Number: 60628-96-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/55/00/28935500.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty