Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorostyrene ≥96.0% (by GC) stabilized

Supplier: TCI America
Synonyms: 1-Chloro-2-vinylbenzene

C0288-5G C0288-25G
TCC0288-5GEA 78.14 USD InStock
TCC0288-5G TCC0288-25G
2-Chlorostyrene ≥96.0% (by GC) stabilized
2-Chlorostyrene
(stabilized with TBC)
CAS Number: 2039-87-4
MDL Number: MFCD00000567
Molecular Formula: C8H7Cl
Molecular Weight: 138.59
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 187
Melting point (°C): -63
Flash Point (°C): 58
Specific Gravity (20/20): 1.10
Storage Temperature: 0-10°C


Formula: C₈H₇Cl
MW: 138.6 g/mol
Boiling Pt: 187 °C
Melting Pt: –63 °C
Density: 1.10 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00000567
CAS Number: 2039-87-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/73/34/28937334.jpg
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty