Synonyms:
5-Chloropentan-2-one
C0867-25ML
TCC0867-25MLEA
200.97
USD
InStock
TCC0867-25ML
5-Chloro-2-pentanone ≥98.0% (by GC) stabilized
5-Chloro-2-pentanone
(stabilized with K2CO3)
CAS Number: 5891-21-4
MDL Number: MFCD00001008
Molecular Formula: C5H9ClO
Molecular Weight: 120.58
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Flash Point (°C): 35
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: 3-Chloropropyl Methyl Ketone