Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Bromo-2-chlorotoulene ≥98.0% (by GC)

Supplier: TCI America
Synonyms: 2-Chlorobenzyl bromide1-(Bromomethyl)-2-chlorobenzene

C1443-25G C1443-5G
TCC1443-25GEA 113.68 USD InStock
TCC1443-25G TCC1443-5G
α-Bromo-2-chlorotoulene ≥98.0% (by GC)
α-Bromo-2-chlorotoulene
CAS Number: 611-17-6
MDL Number: MFCD00000566
Molecular Formula: C7H6BrCl
Molecular Weight: 205.48
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 109
Specific Gravity (20/20): 1.58

Other Product Names / Synonyms: alpha-Bromo-2-chlorotoluene


Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 130 °C
Density: 1.58 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00000566
CAS Number: 611-17-6
UN: 3265
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/86/68/28938668.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty