Synonyms:
(Carbethoxyethylidene)triphenylphosphorane
, Ethyl 2-(triphenylphosphoranylidene)propanoate
C1641-5G
TCC1641-5GEA
97
USD
InStock
TCC1641-5G
(1-Ethoxycarbonylethylidene)triphenylphosphorane ≥97.0% (by HPLC)
(1-Ethoxycarbonylethylidene)triphenylphosphorane
CAS Number: 5717-37-3
MDL Number: MFCD00009160
Molecular Formula: C23H23O2P
Molecular Weight: 362.41
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 161
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (1-Carbethoxyethylidene)triphenylphosphorane, (1-Ethoxycarbonylethylidene)triphenylphosphorane, 2-(Triphenylphosphoranylidene)propionic Acid Ethyl Ester