Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrachloro-p-benzoquinone ∼2% in DMF

Supplier: TCI America
Synonyms: Tetrachloro-1,4-benzoquinone Tetrachloroquinone 4-Chloranil p-Chloranil 2,3,5,6-Tetrachloro-1,4-benzoquinone

C1770-10ML
TCC1770-10MLEA 45.68 USD InStock
TCC1770-10ML
Tetrachloro-p-benzoquinone ∼2% in DMF
Tetrachloro-p-benzoquinone
[for Detection of Primary and Secondary Amines]
CAS Number: 118-75-2
MDL Number: MFCD00001594
Molecular Formula: C6Cl4O2
Molecular Weight: 245.86
Form: Clear Liquid
Color: Yellow
Specific Gravity (20/20): 0.96

Other Product Names / Synonyms: Tetrachloro-p-benzoquinone


Formula: C₆Cl₄O₂
MW: 245.88 g/mol
Density: 0.96 (20 °C)
MDL Number: MFCD00001594
CAS Number: 118-75-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/59/28938959.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty