Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorothiophenol ≥97.0% (by GC)

Supplier: TCI America
Synonyms: o-Chlorothiophenol1-Chloro-2-mercaptobenzene2-Chlorobenzene-1-thiol2-Chlorobenzenethiol2-chlorothiophenol

C1975-25G
TCC1975-25GEA 132.97 USD InStock
TCC1975-25G
2-Chlorothiophenol ≥97.0% (by GC)
2-Chlorothiophenol
CAS Number: 6320-03-2
Molecular Formula: C6H5ClS
Molecular Weight: 144.62
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 206
Flash Point (°C): 88
Specific Gravity (20/20): 1.27

Other Product Names / Synonyms: 1-Chloro-2-mercaptobenzene, 2-Chlorothiophenol


Formula: C₆H₅ClS
MW: 144.62 g/mol
Boiling Pt: 206 °C
Density: 1.27 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00004830
CAS Number: 6320-03-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/90/48/28939048.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty