Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloropiperidine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C2182-5G C2182-25G
TCC2182-5GEA 104.84 USD InStock
TCC2182-5G TCC2182-25G
4-Chloropiperidine ≥97.0% (by GC, titration analysis)
4-Chloropiperidine
CAS Number: 5382-18-3
Molecular Formula: C5H10ClN
Molecular Weight: 119.59
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 48
Specific Gravity (20/20): 1.09
Storage Temperature: 0-10°C


Formula: C₅H₁₀ClN
Boiling Pt: 60 °C
Density: 1.09 (20 °C)
Flash Pt: 48 °C
CAS Number: 5382-18-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/92/01/28939201.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty