Synonyms:
H-Cyclohexyl-Gly-OH(S)-alpha-Aminocyclohexylacetic AcidH-Chg-OH
C2569-5G
C2569-1G
TCC2569-1GEA
41.62
USD
InStock
TCC2569-1G
TCC2569-5G
(S)-2-Amino-2-cyclohexylacetic acid ≥98.0% (by HPLC, titration analysis)
(S)-2-Amino-2-cyclohexylacetic acid
CAS Number: 14328-51-9
MDL Number: MFCD01311679
Molecular Formula: C8H15NO2
Molecular Weight: 157.21
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 255
Specific rotation [a]20/D: 30 deg (C=1, 1ml/L HCl)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-alpha-Aminocyclohexylacetic Acid, H-Chg-OH