Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloro-m-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2576-5G
TCC2576-5GEA 83 USD InStock
TCC2576-5G
6-Chloro-m-toluidine ≥98.0% (by GC, titration analysis)
6-Chloro-m-toluidine
CAS Number: 95-81-8
MDL Number: MFCD00007674
Molecular Formula: C7H8ClN
Molecular Weight: 141.60
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 230
Flash Point (°C): 107
Freezing point (°C): 29

Other Product Names / Synonyms: 3-Amino-4-chlorotoluene, 6-Chloro-m-toluidine


Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 230 °C
Flash Pt: 107 °C
MDL Number: MFCD00007674
CAS Number: 95-81-8
UN: 2239
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/95/75/28939575.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty