Synonyms:
1-(2-Chlorotrityl)imidazole1-[(2-Chlorophenyl)diphenylmethyl]imidazole
C2867-25G
C2867-5G
TCC2867-25GEA
169.46
USD
InStock
TCC2867-25G
TCC2867-5G
Clotrimazole ≥98.0% (by HPLC, titration analysis)
Clotrimazole
CAS Number: 23593-75-1
MDL Number: MFCD00057220
Molecular Formula: C22H17ClN2
Molecular Weight: 344.84
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 147
Lambda max.: 556 nm (Phosphate buffer sol.)
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antifungal Ingredients
Other Product Names / Synonyms: 1-(2-Chlorotrityl)imidazole, 1-[(2-Chlorophenyl)diphenylmethyl]imidazole