Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chlorobenzofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: 5-Chlorobenzo[b]furan-2-carboxylic acid

C2890-5G C2890-1G
TCC2890-1GEA 106 USD InStock
TCC2890-1G TCC2890-5G
5-Chlorobenzofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)
5-Chlorobenzofuran-2-carboxylic acid
CAS Number: 10242-10-1
MDL Number: MFCD00060512
Molecular Formula: C9H5ClO3
Molecular Weight: 196.59
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 266
Lambda max.: 301 nm (EtOH)

Other Product Names / Synonyms: 5-Chlorocoumarilic Acid


https://us-legacy.vwr.com/stibo/web/std.lang.all/69/72/28966972.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty