Synonyms:
Allyl disulphide
, Diallyldisulfide
, Diallyldisulphide
, Allyl disulfide
D0071-25G
TCD0071-25GEA
223.3
USD
InStock
TCD0071-25G
Diallyl disulfide ≥75.0% (by GC)
Diallyl disulphide
CAS Number: 2179-57-9
MDL Number: MFCD00008656
Molecular Formula: C6H10S2
Molecular Weight: 146.27
Purity/Analysis Method: 80.0% (GC)
Form: Clear Liquid
Flash Point (°C): 62
Specific Gravity (20/20): 1.02
Storage Temperature: 0-10°C