D0380-25G
TCD0380-25GEA
37.5
USD
InStock
TCD0380-25G
Dichloro(methyl)silane ≥98.0% (by GC, titration analysis)
Dichloro(methyl)silane
CAS Number: 75-54-7
MDL Number: MFCD00000494
Molecular Formula: CH4Cl2Si
Molecular Weight: 115.03
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 41
Flash Point (°C): -20
Specific Gravity (20/20): 1.12
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Methyldichlorosilane