Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dichloroacetate ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Sodium 2,2-dichloroacetate

D1048-25G
TCD1048-25GEA 77.14 USD InStock
TCD1048-25G
Sodium dichloroacetate ≥97.0% (by titrimetric analysis)
Sodium dichloroacetate
CAS Number: 2156-56-1
MDL Number: MFCD00070489
Molecular Formula: C2H2Cl2O2
Molecular Weight: 150.92
Purity/Analysis Method: 97.0% (T)
Form: Crystal

Other Product Names / Synonyms: Dichloroacetic Acid Sodium Salt


Formula: C₂HCl₂NaO₂
MW: 150.92 g/mol
MDL Number: MFCD00070489
CAS Number: 2156-56-1
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty