Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Dinitrotoluene ≥99.0% (by GC)

Supplier: TCI America

D1151-25G
TCD1151-25GEA 57.83 USD InStock
TCD1151-25G
2,6-Dinitrotoluene ≥99.0% (by GC)
2,6-Dinitrotoluene
CAS Number: 606-20-2
MDL Number: MFCD00007158
Molecular Formula: C7H6N2O4
Molecular Weight: 182.14
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 65
Flash Point (°C): 207


Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Melting Pt: 65 °C
Flash Pt: 207 °C
MDL Number: MFCD00007158
CAS Number: 606-20-2
UN: 3454
ADR: 6.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/80/85/28978085.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty