Synonyms:
(1,1-Dimethoxyethyl)dimethylamine
, 1,1-Dimethoxy-N,N-dimethylethanamine
D1652-25ML
TCD1652-25MLEA
102.13
USD
InStock
TCD1652-25ML
N,N-Dimethylacetamide dimethyl acetal ≥90.0% (by titrimetric analysis) stabilized
N,N-Dimethylacetamide dimethyl acetal
(stabilized with 5-10% Methanol)
CAS Number: 18871-66-4
MDL Number: MFCD00008476
Molecular Formula: C6H15NO2
Molecular Weight: 133.19
Purity/Analysis Method: 90.0% (T)
Form: Clear Liquid
Flash Point (°C): 8
Specific Gravity (20/20): 0.91
Other Product Names / Synonyms: N-(1,1-Dimethoxyethyl)dimethylamine