Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichlorodiphenylgermane ≥95.0% (by GC)

Supplier: TCI America
Synonyms: Diphenylgermanium dichlorideDichlorodiphenylgermane

D1824-1G
TCD1824-1GEA 99 USD InStock
TCD1824-1G
Dichlorodiphenylgermane ≥95.0% (by GC)
Dichlorodiphenylgermane
CAS Number: 1613-66-7
MDL Number: MFCD00013587
Molecular Formula: C12H10Cl2Ge
Molecular Weight: 297.74
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Melting point (°C): 9
Flash Point (°C): 160
Specific Gravity (20/20): 1.43

Other Product Names / Synonyms: Diphenylgermanium Dichloride


Formula: C₁₂H₁₀Cl₂Ge
Melting Pt: 9 °C
Density: 1.43 (20 °C)
Flash Pt: 160 °C
MDL Number: MFCD00013587
CAS Number: 1613-66-7
UN: 3265
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/86/09/28978609.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty