Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dess-Martin periodinane ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: DMP(1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-oneAcetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester

D2045-25G D2045-5G D2045-1G
TCD2045-1GEA 38.71 USD InStock
TCD2045-1G TCD2045-25G TCD2045-5G
Dess-Martin periodinane ≥95.0% (by titrimetric analysis)
Dess-Martin periodinane
CAS Number: 87413-09-0
MDL Number: MFCD00130127
Molecular Formula: C13H13IO8
Molecular Weight: 424.14
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 124
Storage Temperature: 0°C

Product Categories: Synthetic Organic Chemistry, Hypervalent Iodine Compounds, Oxidation
Other Product Names / Synonyms: 1,1,1-Triacetoxy-1,1-dihydro-1,2-benziodoxol-3(1H)-one


Formula: C13H13IO8
MW: 424.15 g/mol
Melting Pt: 124 °C
MDL Number: MFCD00130127
CAS Number: 87413-09-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/36/28978836.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty