Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Dichloro-3-nitrotoluene ≥98.0% (by GC)

Supplier: TCI America
Synonyms: 1,3-Dichloro-2-methyl-4-nitrobenzene

D2106-25G
TCD2106-25GEA 212 USD InStock
TCD2106-25G
2,6-Dichloro-3-nitrotoluene ≥98.0% (by GC)
2,6-Dichloro-3-nitrotoluene
CAS Number: 29682-46-0
MDL Number: MFCD00024185
Molecular Formula: C7H5Cl2NO2
Molecular Weight: 206.02
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 55
Flash Point (°C): 110

Other Product Names / Synonyms: 1,3-Dichloro-2-methyl-4-nitrobenzene


Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Melting Pt: 55 °C
Flash Pt: 110 °C
MDL Number: MFCD00024185
CAS Number: 29682-46-0
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty