Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenyliodonium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D2356-25G D2356-5G
TCD2356-25GEA 652.65 USD InStock
TCD2356-25G TCD2356-5G
Diphenyliodonium chloride ≥98.0% (by titrimetric analysis)
Diphenyliodonium chloride
CAS Number: 1483-72-3
MDL Number: MFCD00011909
Molecular Formula: C12H10ClI
Molecular Weight: 316.57
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Formula: C₁₂H₁₀ClI
MW: 316.57 g/mol
MDL Number: MFCD00011909
CAS Number: 1483-72-3
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/92/87/28979287.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty