Synonyms:
alpha,3,5-Tribromotoluene
, 3,5-Dibromobenzyl bromide
, 1,3-Dibromo-5-(bromomethyl)benzene
D4020-5G
D4020-25G
TCD4020-25GEA
239.54
USD
InStock
TCD4020-25G
TCD4020-5G
α,3,5-Tribromotoluene ≥95.0% (by GC)
α,3,5-Tribromotoluene
CAS Number: 56908-88-4
MDL Number: MFCD00052415
Molecular Formula: C7H5Br3
Molecular Weight: 328.83
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Boiling point (°C): 169
Melting point (°C): 94
Other Product Names / Synonyms: alpha,3,5-Tribromotoluene