Synonyms:
2,3-Naphthalenediamine 2,3-naphthylenediamine DAN
D4101-100MG
TCD4101-100MGEA
58.06
USD
InStock
TCD4101-100MG
2,3-Diaminonaphthalene ≥98.0% (by GC, titration analysis) for biochemical research
2,3-Diaminonaphthalene
[for Biochemical Research]
CAS Number: 771-97-1
MDL Number: MFCD00004116
Molecular Formula: C10H10N2
Molecular Weight: 158.20
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 198
Storage Temperature: 0-10°C
Product Categories: Bioscience, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: 2,3-Naphthalenediamine