Synonyms:
5-Chloro-1-[1-[3-(2-oxo-1-benzimidazolyl)propyl]-4-piperidyl]benzimidazol-2-one
D4125-200MG
D4125-1G
TCD4125-200MGEA
30.45
USD
InStock
TCD4125-200MG
TCD4125-1G
Domperidone ≥98.0% (by HPLC, titration analysis)
Domperidone
CAS Number: 57808-66-9
MDL Number: MFCD00069256
Molecular Formula: C22H24ClN5O2
Molecular Weight: 425.92
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 243
Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 5-Chloro-1-[1-[3-(2-oxo-1-benzimidazolyl)propyl]-4-piperidyl]benzimidazol-2-one