Synonyms:
(2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol
D4137-5G
TCD4137-5GEA
132.97
USD
InStock
TCD4137-5G
2'-Deoxyadenosine, anhydrous ≥98.0% (by HPLC, titration analysis)
2'-Deoxyadenosine
CAS Number: 958-09-8
MDL Number: MFCD00005754
Molecular Formula: C10H13N5O3
Molecular Weight: 251.25
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 191
Specific rotation [a]20/D: -27 deg (C=0.5, H2O)
Storage Temperature: 0-10°C