D4336-1G
TCD4336-1GEA
60
USD
InStock
TCD4336-1G
Dioctyldichlorosilane ≥96.0% (by GC)
Dioctyldichlorosilane
CAS Number: 18416-07-4
MDL Number: MFCD01074472
Molecular Formula: C16H34Cl2Si
Molecular Weight: 325.43
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Specific Gravity (20/20): 0.94