Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Synonyms: N-(tert-Butoxycarbonyl)-L-glutamic Acid Dimethyl EsterDimethyl N-Boc-L-glutamateN-Boc-L-glutamic Acid Dimethyl EsterBoc-Glu(OMe)-OMe

D4536-1G D4536-5G
TCD4536-1GEA 37 USD InStock
TCD4536-1G TCD4536-5G
Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate ≥98.0% (by HPLC, total nitrogen)
Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate
CAS Number: 59279-60-6
MDL Number: MFCD03094774
Molecular Formula: C12H21NO6
Molecular Weight: 275.30
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 45
Specific rotation [a]20/D: 14 deg (C=2, CHCl3)

Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-(tert-Butoxycarbonyl)-L-glutamic Acid Dimethyl Ester, Dimethyl N-Boc-L-glutamate, N-Boc-L-glutamic Acid Dimethyl Ester, Boc-Glu(OMe)-OMe


Formula: C₁₂H₂₁NO₆
MW: 275.30 g/mol
Melting Pt: 45 °C
MDL Number: MFCD03094774
CAS Number: 59279-60-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/66/62/28986662.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty