Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Diphenylphosphino)biphenyl ≥98.0% (by GC)

Supplier: TCI America

D4688-1G
TCD4688-1GEA 30.45 USD InStock
TCD4688-1G
2-(Diphenylphosphino)biphenyl ≥98.0% (by GC)
2-(Diphenylphosphino)biphenyl
CAS Number: 13885-09-1
MDL Number: MFCD11559063
Molecular Formula: C24H19P
Molecular Weight: 338.39
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 82

Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: (2-Biphenylyl)diphenylphosphine


Formula: C24H19P
MW: 338.38 g/mol
Melting Pt: 82 °C
MDL Number: MFCD11559063
CAS Number: 13885-09-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/68/22/28986822.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty