Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Chloro-2-fluorotoulene ≥98.0% (by GC)

Supplier: TCI America
Synonyms: 1-(Chloromethyl)-2-fluorobenzene 2-Fluorobenzyl chloride α-Chloro-o-fluorotoulene

F0211-25G F0211-5G
TCF0211-5GEA 16.24 USD InStock
TCF0211-5G TCF0211-25G
α-Chloro-2-fluorotoulene ≥98.0% (by GC)
α-Chloro-2-fluorotoulene
CAS Number: 345-35-7
MDL Number: MFCD00000892
Molecular Formula: C7H6ClF
Molecular Weight: 144.57
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 57
Specific Gravity (20/20): 1.22


Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 175 °C
Density: 1.22 (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00000892
CAS Number: 345-35-7
UN: 2920
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/17/28968817.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty