Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Fluoro-o-toluidine ≥98.0% (by GC)

Supplier: TCI America
Synonyms: 2-Amino-4-fluorotoluene 5-Fluoro-2-methylaniline

F0232-25G F0232-5G
TCF0232-5GEA 34 USD InStock
TCF0232-5G TCF0232-25G
5-Fluoro-o-toluidine ≥98.0% (by GC)
5-Fluoro-o-toluidine
CAS Number: 367-29-3
MDL Number: MFCD00007764
Molecular Formula: C7H8FN
Molecular Weight: 125.15
Purity/Analysis Method: 98.0% (GC)
Boiling point (°C): 83
Flash Point (°C): 89
Freezing point (°C): 36

Other Product Names / Synonyms: 2-Amino-4-fluorotoluene, 5-Fluoro-o-toluidine


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 83 °C
Flash Pt: 89 °C
MDL Number: MFCD00007764
CAS Number: 367-29-3
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/35/28968835.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty