Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorobenzylamine ≥98.0% (by GC)

Supplier: TCI America
Synonyms: (4-Fluorophenyl)methanamine

F0252-25G
TCF0252-25GEA 56.82 USD InStock
TCF0252-25G
4-Fluorobenzylamine ≥98.0% (by GC)
4-Fluorobenzylamine
CAS Number: 140-75-0
MDL Number: MFCD00008120
Molecular Formula: C7H8FN
Molecular Weight: 125.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Flash Point (°C): 66
Specific Gravity (20/20): 1.10


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 183 °C
Density: 1.10 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00008120
CAS Number: 140-75-0
UN: 2735
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/51/28968851.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty