Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formyltetrathiafulvalene ≥98.0% (by GC)

Supplier: TCI America
Synonyms: Formyl-TTF[2,2'-Bi(1,3-dithiolylidene)]-4-carboxaldehyde

F0285-1G
TCF0285-1GEA 502.21 USD InStock
TCF0285-1G
Formyltetrathiafulvalene ≥98.0% (by GC)
Formyltetrathiafulvalene
CAS Number: 68128-94-9
Molecular Formula: C7H4OS4
Molecular Weight: 232.35
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 113

Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: Formyl-TTF, [2,2'-Bi(1,3-dithiolylidene)]-4-carboxaldehyde


Formula: C₇H₄OS₄
MW: 232.35 g/mol
Melting Pt: 113 °C
CAS Number: 68128-94-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/81/28968881.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty